Assay Details
Target Gene Details
Entrez Gene ID: | 16333 |
Gene Name: | insulin I |
Gene Aliases: |
Ins-1, Ins2-rs1 |
Location: |
Chr.19:52264323-52265476 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ins1 | NM_008386.3 | 2 | 747 | NP_032412.3 |
| BP771435.1 | 1 | 316 |
More Information
Set Membership: |
|

Genomic Map