Assay Details
Target Gene Details
Entrez Gene ID: | 72937 |
Gene Name: | RIKEN cDNA 2900008C10 gene |
Gene Aliases: |
- |
Location: |
Chr.X:12134454-12160346 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 2900008C10Rik | NR_045434.1 | 4 | 1291 | |
| AK013498.1 | 2 | 717 | ||
| BP776822.1 | 1 | 115 |
Target Gene Details
Entrez Gene ID: | 71458 |
Gene Name: | BCL6 interacting corepressor |
Gene Aliases: |
5830466J11Rik, 8430401K06Rik, BcorR, D930024N20Rik, mKIAA1575 |
Location: |
Chr.X:12036737-12160355 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bcor | NM_001168321.1 | NP_001161793.1 | ||
| AK129398.1 | BAC98208.1 |
More Information
Set Membership: |
|

Genomic Map