Assay Details
Target Gene Details
Entrez Gene ID: | 56736 |
Gene Name: | ring finger protein 14 |
Gene Aliases: |
2310075C09Rik, 2610005D23Rik, AA986456, AU041447, D18Ertd188e, D7Bwg0165e, Triad2 |
Location: |
Chr.18:38296635-38317852 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rnf14 | NM_001164621.1 | 10 | 2460 | NP_001158093.1 |
| NM_001164622.1 | 8 | 1995 | NP_001158094.1 | |
| NM_020012.2 | 9 | 2147 | NP_064396.2 | |
| AF249667.1 | 9 | 2093 | AAF74266.1 | |
| AK009688.1 | 4 | 758 | ||
| AK011316.2 | 10 | 2148 | BAB27541.1 | |
| AK038986.1 | 8 | 1937 | ||
| BC054841.1 | 9 | 2077 | AAH54841.1 | |
| BC094250.1 | 9 | 2098 | AAH94250.1 |
More Information
Set Membership: |
|

Genomic Map