Assay Details
Target Gene Details
Entrez Gene ID: | 225207 |
Gene Name: | zinc finger protein 521 |
Gene Aliases: |
B930086A16Rik, Evi3, Znf521 |
Location: |
Chr.18:13687013-13972787 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfp521 | NM_145492.4 | 8 | 4677 | NP_663467.1 |
| AK052655.1 | 8 | 4581 | ||
| AK077697.1 | 5 | 1499 | BAC36964.1 | |
| AK081096.1 | 6 | 2131 | BAC38132.1 | |
| AK131130.1 | 7 | 4762 | BAD21380.1 | |
| AY147406.1 | 8 | 4580 | AAN39839.1 | |
| BC021376.1 | 8 | 4493 | AAH21376.1 |
More Information
Set Membership: |
|

Genomic Map