Assay Details
Target Gene Details
Entrez Gene ID: | 171170 |
Gene Name: | muscleblind-like 3 (Drosophila) |
Gene Aliases: |
A530038J18Rik, AI661274, CHCR, E430034C16Rik, MBLX, MBLX39, MBXL |
Location: |
Chr.X:51113494-51206035 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mbnl3 | NM_134163.4 | 8 | 6161 | NP_598924.1 |
| AK040889.1 | 1 | 1980 | ||
| AK053108.1 | 1 | 723 |
More Information
Set Membership: |
|

Genomic Map