Assay Details
Target Gene Details
Entrez Gene ID: | 108101 |
Gene Name: | fermitin family homolog 3 (Drosophila) |
Gene Aliases: |
C79673, Kindlin3 |
Location: |
Chr.19:6998958-7019469 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fermt3 | NM_153795.2 | 15 | 2128 | NP_722490.1 |
| AK090332.1 | 15 | 2025 | ||
| AK142912.1 | 15 | 2129 | ||
| AK149737.1 | 14 | 2551 | ||
| AK158357.1 | 15 | 2025 | ||
| AK169682.1 | 15 | 2039 | BAE41302.1 | |
| BC025119.1 | 15 | 2060 | AAH25119.1 | |
| BC032204.1 | 15 | 2104 | AAH32204.1 | |
| BC066803.1 | 14 | 2410 | AAH66803.1 |

Genomic Map