Assay Details
Target Gene Details
Entrez Gene ID: | 240660 |
Gene Name: | solute carrier family 35, member G1 |
Gene Aliases: |
AA590464, D330039I19Rik, Tmem20 |
Location: |
Chr.19:38395980-38405607 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc35g1 | NM_175507.3 | 3 | 991 | NP_780716.1 |
| AK041618.1 | 3 | 991 | BAC31006.1 | |
| AK052365.1 | 3 | 895 | ||
| AK171987.1 | 3 | 923 | BAE42760.1 | |
| BC137663.1 | 3 | 1002 | ||
| BC137664.1 | 3 | 1002 |
More Information
Set Membership: |
|

Genomic Map