Assay Details
Target Gene Details
Entrez Gene ID: | 54646 |
Gene Name: | protein phosphatase 1, regulatory (inhibitor) subunit 3F |
Gene Aliases: |
DXImx48e, R3F, RF3, Sfc15 |
Location: |
Chr.X:7558562-7574281 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppp1r3f | NM_001290574.1 | 1 | 451 | NP_001277503.1 |
| NM_138605.3 | 1 | 451 | NP_613071.3 | |
| AF229644.1 | 1 | 290 | AAF66954.2 | |
| BC059275.1 | 1 | 385 | AAH59275.1 | |
| BC138137.1 | 1 | 283 | ||
| BC138138.1 | 1 | 283 | ||
| BY727062.1 | 1 | 233 |
Target Gene Details
Entrez Gene ID: | 78185 |
Gene Name: | protein phosphatase 1, regulatory subunit 3F, opposite strand |
Gene Aliases: |
4930524L23Rik |
Location: |
Chr.X:7573600-7581016 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppp1r3fos | NR_029473.1 | 1 | 232 | |
| AK048169.1 | 1 | 232 | ||
| AK080141.1 | BAC37832.1 | |||
| AK080337.1 | 1 | 37 |
More Information
Set Membership: |
|

Genomic Map