Assay Details
Target Gene Details
Entrez Gene ID: | 54646 |
Gene Name: | protein phosphatase 1, regulatory (inhibitor) subunit 3F |
Gene Aliases: |
DXImx48e, R3F, RF3, Sfc15 |
Location: |
Chr.X:7558562-7574281 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppp1r3f | NM_001290574.1 | 4 | 2717 | NP_001277503.1 |
| NM_138605.3 | 4 | 2714 | NP_613071.3 | |
| AF229644.1 | 4 | 2553 | AAF66954.2 | |
| BC059275.1 | 4 | 2651 | AAH59275.1 | |
| BC138137.1 | 4 | 2546 | ||
| BC138138.1 | 4 | 2546 |
More Information
Set Membership: |
|

Genomic Map