Assay Details
Target Gene Details
Entrez Gene ID: | 14585 |
Gene Name: | glial cell line derived neurotrophic factor family receptor alpha 1 |
Gene Aliases: |
AU042498 |
Location: |
Chr.19:58235581-58455946 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gfra1 | NM_001285457.1 | 4 | 949 | NP_001272386.1 |
| NM_010279.3 | 6 | 1165 | NP_034409.1 | |
| NR_104342.1 | 6 | 1165 | ||
| AB000800.1 | 5 | 719 | BAA19185.1 | |
| AF012811.1 | AAB65855.1 | |||
| AF015172.1 | 4 | 932 | AAB86600.1 | |
| AK076171.1 | 6 | 1085 | ||
| AK139987.1 | ||||
| BC054378.1 | 5 | 836 | AAH54378.1 | |
| KM079499.1 | 5 | 824 |
Target Gene Details
Entrez Gene ID: | 791347 |
Gene Name: | predicted gene 10007 |
Gene Aliases: |
ENSMUSG00000056739 |
Location: |
Chr.19:58296204-58300348 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm10007 | NR_040449.1 | |||
| BY300051.1 | ||||
| CJ183106.1 |
More Information
Set Membership: |
|

Genomic Map