Assay Details
Target Gene Details
Entrez Gene ID: | 225875 |
Gene Name: | leucine rich repeat and fibronectin type III domain containing 4 |
Gene Aliases: |
BC023156, Salm3 |
Location: |
Chr.19:4611784-4615667 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lrfn4 | NM_153388.4 | 3 | 2779 | NP_700437.2 |
| AI415633.1 | 1 | 175 | ||
| AK081560.1 | 2 | 3062 | BAC38259.1 | |
| AK163603.1 | 3 | 2637 | BAE37415.1 | |
| AK205631.1 | 1 | 177 | ||
| AK207195.1 | ||||
| BC023036.1 | 2 | 2392 | AAH23036.1 | |
| BC023156.1 | 3 | 2435 | AAH23156.1 |
Target Gene Details
Entrez Gene ID: | 18563 |
Gene Name: | pyruvate carboxylase |
Gene Aliases: |
Pc, Pcb |
Location: |
Chr.19:4510472-4621752 on Build GRCm38 |
Assay Gene Location: | Within Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pcx | NM_001162946.1 | NP_001156418.1 | ||
| NM_008797.3 | NP_032823.2 | |||
| AK077665.1 | BAC36939.1 | |||
| AK148328.1 | BAE28484.1 | |||
| AK170596.1 | BAE41902.1 | |||
| AK170871.1 | ||||
| AK172320.1 | BAE42943.1 | |||
| BC055030.1 | AAH55030.1 | |||
| L09192.1 | AAA39737.1 | |||
| M97957.1 | AAA39897.1 |
More Information
Set Membership: |
|

Genomic Map