Assay Details
Target Gene Details
Entrez Gene ID: | 209446 |
Gene Name: | transcription factor E3 |
Gene Aliases: |
F830016E06Rik, Tcfe3, Tfe-3, bHLHe33 |
Location: |
Chr.X:7762538-7775202 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tfe3 | NM_001105196.1 | 1 | 133 | NP_001098666.1 |
| NM_001271489.1 | 1 | 133 | NP_001258418.1 | |
| NM_001271490.1 | 1 | 133 | NP_001258419.1 | |
| NM_172472.3 | 1 | 133 | NP_766060.2 | |
| AF510653.1 | AAM95608.1 | |||
| AK089762.1 | 1 | 79 | BAC40955.1 | |
| AK139756.1 | 1 | 130 | BAE24128.1 | |
| AK144657.1 | 1 | 98 | BAE25994.1 | |
| AK144751.1 | 1 | 119 | ||
| AK145029.1 | 1 | 99 | ||
| AK145832.1 | 1 | 117 | ||
| AK154690.1 | 1 | 99 | ||
| AK159162.1 | 1 | 33 | ||
| BC032963.1 | ||||
| BC056358.1 | 1 | 121 | AAH56358.1 | |
| BC063047.1 | 1 | 121 | AAH63047.1 | |
| BY299767.1 | 1 | 134 |

Genomic Map