Assay Details
Target Gene Details
Entrez Gene ID: | 209446 |
Gene Name: | transcription factor E3 |
Gene Aliases: |
F830016E06Rik, Tcfe3, Tfe-3, bHLHe33 |
Location: |
Chr.X:7762538-7775202 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tfe3 | NM_001105196.1 | 3 | 665 | NP_001098666.1 |
| NM_001105197.1 | 3 | 351 | NP_001098667.1 | |
| NM_001271489.1 | 3 | 646 | NP_001258418.1 | |
| NM_001271490.1 | 3 | 818 | NP_001258419.1 | |
| NM_001271491.1 | 3 | 385 | NP_001258420.1 | |
| NM_172472.3 | 3 | 665 | NP_766060.2 | |
| AF510653.1 | 3 | 507 | AAM95608.1 | |
| AK089762.1 | 2 | 427 | BAC40955.1 | |
| AK139756.1 | 3 | 662 | BAE24128.1 | |
| AK144657.1 | 3 | 630 | BAE25994.1 | |
| AK144751.1 | 3 | 632 | ||
| AK145029.1 | 3 | 784 | ||
| AK145832.1 | 3 | 630 | ||
| AK147405.1 | 2 | 1120 | ||
| AK150372.1 | 3 | 418 | ||
| AK151447.1 | 3 | 336 | ||
| AK152221.1 | 3 | 336 | BAE31048.1 | |
| AK153051.1 | 3 | 418 | ||
| AK154690.1 | 3 | 631 | ||
| AK155883.1 | 3 | 387 | BAE33481.1 | |
| AK159162.1 | 3 | 546 | ||
| AK190908.1 | ||||
| BC032963.1 | ||||
| BC056358.1 | 3 | 653 | AAH56358.1 | |
| BC063047.1 | 3 | 653 | AAH63047.1 | |
| BY208269.1 | 3 | 321 | ||
| S76673.1 | AAB21130.1 |

Genomic Map