Assay Details
Target Gene Details
Entrez Gene ID: | 67843 |
Gene Name: | solute carrier family 35, member A4 |
Gene Aliases: |
2610030J16Rik |
Location: |
Chr.18:36679215-36683862 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc35a4 | NM_001083317.1 | 1 | 342 | NP_001076786.1 |
| NM_026404.2 | 1 | 342 | NP_080680.2 | |
| AK011633.1 | BAB27747.1 | |||
| AK018546.1 | BAB31267.1 | |||
| AK031132.1 | BAC27268.1 | |||
| AK139953.1 | ||||
| AK145812.1 | BAE26666.1 | |||
| AK164797.1 | BAE37923.1 | |||
| AK170045.1 | BAE41529.1 | |||
| BY088803.1 | ||||
| BY732053.1 | 1 | 321 |
More Information
Set Membership: |
|

Genomic Map