Assay Details
Target Gene Details
Entrez Gene ID: | 72065 |
Gene Name: | RAP2C, member of RAS oncogene family |
Gene Aliases: |
2010200P20Rik, AI194294, AL022976 |
Location: |
Chr.X:51003912-51020092 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rap2c | NM_172413.2 | 1 | 133 | NP_766001.1 |
| AK087971.1 | 1 | 80 | BAC40066.1 | |
| AK146057.1 | 1 | 112 | BAE26866.1 | |
| AK146956.1 | 1 | 175 | BAE27564.1 | |
| AK147015.1 | 1 | 175 | BAE27609.1 | |
| AK179813.1 | 1 | 66 | ||
| BC027363.1 | 1 | 97 | AAH27363.3 | |
| BC050056.1 | 1 | 95 | ||
| BC064814.1 | 1 | 129 | AAH64814.2 | |
| BC094890.1 | 1 | 133 | AAH94890.1 |
More Information
Set Membership: |
|

Genomic Map