Assay Details
Target Gene Details
Entrez Gene ID: | 14266 |
Gene Name: | AF4/FMR2 family, member 2 |
Gene Aliases: |
FMR2P, Fmr2, Ox19, Oxh |
Location: |
Chr.X:69360331-69872054 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Aff2 | NM_008032.3 | 3 | 921 | NP_032058.2 |
| AJ001549.1 | 3 | 925 | CAA04821.1 | |
| AJ748649.1 | 1 | 76 | CAG38683.1 |
More Information
Set Membership: |
|

Genomic Map