Assay Details
Target Gene Details
Entrez Gene ID: | 107765 |
Gene Name: | ankyrin repeat domain 1 (cardiac muscle) |
Gene Aliases: |
Alrp, CARP, Crap, MARP1 |
Location: |
Chr.19:36111965-36119844 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ankrd1 | NM_013468.3 | 9 | 1048 | NP_038496.2 |
| AF041847.1 | AAC03533.1 | |||
| AK009655.1 | 9 | 1041 | BAB26419.1 | |
| AK009959.1 | 9 | 1047 | BAB26611.1 | |
| AK145944.1 | 9 | 1089 | BAE26773.1 | |
| AK146471.1 | 9 | 1045 | BAE27197.1 | |
| AK146627.1 | 9 | 1047 | BAE27316.1 | |
| AK146940.1 | 9 | 1045 | BAE27549.1 | |
| BC037138.1 | 9 | 1048 | AAH37138.1 |

Genomic Map