Assay Details
Target Gene Details
Entrez Gene ID: | 83410 |
Gene Name: | cleavage stimulation factor, 3' pre-RNA subunit 2, tau |
Gene Aliases: |
64kDa, C77975, tCstF-64, tauCstF-64 |
Location: |
Chr.19:31082841-31086592 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cstf2t | NM_031249.2 | 1 | 3129 | NP_112539.2 |
| AF322194.1 | 1 | 2980 | AAG40327.1 | |
| AK030121.1 | 1 | 2967 | ||
| AK034853.1 | 1 | 2967 | BAC28855.1 | |
| AK050407.1 | 1 | 3085 | BAC34240.1 | |
| AK082910.1 | 1 | 3001 | BAC38683.1 | |
| AK084696.1 | 1 | 2960 | BAC39256.1 | |
| AK173002.1 | 1 | 2939 | BAD32280.1 | |
| BC026995.1 | 1 | 2937 | AAH26995.1 |
Target Gene Details
Entrez Gene ID: | 19091 |
Gene Name: | protein kinase, cGMP-dependent, type I |
Gene Aliases: |
AW125416, CGKI, Gm19690, Prkg1b, Prkgr1b |
Location: |
Chr.19:30564487-31765304 on Build GRCm38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prkg1 | NM_001013833.3 | NP_001013855.1 | ||
| NM_011160.3 | NP_035290.1 | |||
| AF084547.1 | AAD16044.1 | |||
| AK083991.1 | BAC39087.1 | |||
| BC113162.1 |
More Information
Set Membership: |
|

Genomic Map