Assay Details
Target Gene Details
Entrez Gene ID: | 71738 |
Gene Name: | MAM domain containing 2 |
Gene Aliases: |
1200015L10Rik, AI591564, mamcan |
Location: |
Chr.19:23302609-23448500 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mamdc2 | NM_174857.3 | 6 | 960 | NP_777282.1 |
| AK004794.1 | ||||
| AK035566.1 | 6 | 961 | BAC29108.1 | |
| AK154219.1 | 6 | 1037 | BAE32442.1 | |
| BC042773.1 | 6 | 989 | AAH42773.1 |

Genomic Map