Assay Details
Target Gene Details
Entrez Gene ID: | 74405 |
Gene Name: | EF-hand domain (C-terminal) containing 2 |
Gene Aliases: |
4933407D04Rik, mRib72-2 |
Location: |
Chr.X:17132049-17320461 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Efhc2 | NM_028916.4 | 5 | 899 | NP_083192.2 |
| AK016714.1 | 5 | 858 | BAB30393.1 | |
| AK029501.1 | 5 | 865 | BAC26479.1 | |
| BC120659.1 | 5 | 834 | ||
| BC125625.1 | 5 | 834 |
More Information
Set Membership: |
|

Genomic Map