Assay Details
Target Gene Details
Entrez Gene ID: | 16151 |
Gene Name: | inhibitor of kappaB kinase gamma |
Gene Aliases: |
1110037D23Rik, AI848108, AI851264, AW124339, IKK[g], NEMO |
Location: |
Chr.X:74393291-74456792 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ikbkg | NM_001136067.2 | 11 | 4132 | NP_001129539.1 |
| NM_001161421.1 | 11 | 4129 | NP_001154893.1 | |
| NM_001161422.1 | 10 | 3950 | NP_001154894.1 | |
| NM_001161423.1 | 10 | 4002 | NP_001154895.1 | |
| NM_001161424.1 | 10 | 3999 | NP_001154896.1 | |
| NM_010547.2 | 10 | 4028 | NP_034677.2 | |
| NM_178590.4 | 10 | 4025 | NP_848705.1 | |
| AK037020.1 | 10 | 4004 | BAC29672.1 |

Genomic Map