Assay Details
Target Gene Details
Entrez Gene ID: | 18563 |
Gene Name: | pyruvate carboxylase |
Gene Aliases: |
Pc, Pcb |
Location: |
Chr.19:4510472-4621752 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pcx | NM_001162946.1 | 14 | 1938 | NP_001156418.1 |
| NM_008797.3 | 13 | 1915 | NP_032823.2 | |
| AK077665.1 | 5 | 743 | BAC36939.1 | |
| AK148328.1 | 14 | 1885 | BAE28484.1 | |
| AK170596.1 | 14 | 1898 | BAE41902.1 | |
| AK170871.1 | 14 | 1888 | ||
| AK172320.1 | 14 | 1866 | BAE42943.1 | |
| BC055030.1 | 14 | 1873 | AAH55030.1 | |
| L09192.1 | 13 | 1891 | AAA39737.1 | |
| M97957.1 | 8 | 1063 | AAA39897.1 |
More Information
Set Membership: |
|

Genomic Map