Assay Details
Target Gene Details
Entrez Gene ID: | 107239 |
Gene Name: | carnosine synthase 1 |
Gene Aliases: |
AI790298, Atpgd1, mKIAA1394 |
Location: |
Chr.19:4164324-4175479 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Carns1 | NM_134148.2 | 5 | 709 | NP_598909.2 |
| AK129347.1 | 5 | 684 | BAC98157.1 |

Genomic Map