Assay Details
Target Gene Details
Entrez Gene ID: | 107239 |
Gene Name: | carnosine synthase 1 |
Gene Aliases: |
AI790298, Atpgd1, mKIAA1394 |
Location: |
Chr.19:4164324-4175479 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Carns1 | NM_134148.2 | 10 | 2475 | NP_598909.2 |
AK129347.1 | 9 | 2632 | BAC98157.1 | |
BC019736.1 | 1 | 197 | AAH19736.1 | |
BC023699.1 | 2 | 889 | AAH23699.1 |
