Assay Details
Target Gene Details
Entrez Gene ID: | 17195 |
Gene Name: | mannose-binding lectin (protein C) 2 |
Gene Aliases: |
L-MBP, MBL, MBL-C, MBP-C |
Location: |
Chr.19:30232906-30239686 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mbl2 | NM_010776.1 | 6 | 804 | NP_034906.1 |
| AK149484.1 | 6 | 834 | BAE28910.1 | |
| BC010760.1 | 6 | 819 | AAH10760.1 | |
| D11440.1 | 6 | 804 | BAA02005.1 | |
| S42294.1 | 6 | 830 | AAB19343.1 |
More Information
Set Membership: |
|

Genomic Map