Assay Details
Target Gene Details
Entrez Gene ID: | 14070 |
Gene Name: | factor 8-associated gene A |
Gene Aliases: |
AI852759, DXUcsf1, F8a1 |
Location: |
Chr.X:73228306-73230795 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| F8a | NM_007978.3 | 1 | 827 | NP_032004.2 |
| AF299331.1 | 1 | 856 | AAG17919.1 | |
| AK033184.1 | 1 | 829 | BAC28188.1 | |
| AK046025.1 | 1 | 842 | ||
| AK046459.1 | 1 | 842 | BAC32739.1 | |
| AK054140.1 | 1 | 828 | BAC35668.1 | |
| AK166334.1 | 1 | 843 | BAE38713.1 | |
| BC131938.1 | 1 | 855 | ||
| BC131940.1 | 1 | 855 | ||
| M83118.1 | 1 | 1115 | AAA37588.1 |

Genomic Map