Assay Details
Target Gene Details
Entrez Gene ID: | 66294 |
Gene Name: | family with sequence similarity 3, member A |
Gene Aliases: |
1810037C20Rik |
Location: |
Chr.X:74384719-74393280 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam3a | NM_025473.3 | 1 | 235 | NP_079749.1 |
| AK007715.1 | 1 | 236 | BAB25208.1 | |
| BC069871.1 | 1 | 370 | AAH69871.1 |
More Information
Set Membership: |
|

Genomic Map