Assay Details
Target Gene Details
Entrez Gene ID: | 14168 |
Gene Name: | fibroblast growth factor 13 |
Gene Aliases: |
Fhf2 |
Location: |
Chr.X:59062139-59585572 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fgf13 | NM_001290414.1 | NP_001277343.1 | ||
| NM_001290415.1 | NP_001277344.1 | |||
| NM_010200.3 | 1 | 467 | NP_034330.2 | |
| AF020737.1 | 1 | 103 | AAB71606.1 | |
| AF199608.1 | AAF31395.1 | |||
| AF199614.1 | AAF31401.1 | |||
| AK141848.1 | BAE24857.1 | |||
| BC018238.1 | 1 | 467 | AAH18238.1 | |
| CJ064772.1 | ||||
| U66202.1 | 1 | 94 | AAB18918.1 |
More Information
Set Membership: |
|

Genomic Map