Assay Details
Target Gene Details
Entrez Gene ID: | 236733 |
Gene Name: | ubiquitin specific peptidase 11 |
Gene Aliases: |
6230415D12Rik, mKIAA4085 |
Location: |
Chr.X:20703909-20720539 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Usp11 | NM_145628.4 | 15 | 2062 | NP_663603.3 |
| AK031767.1 | 15 | 2041 | BAC27541.1 | |
| AK169116.1 | 7 | 885 | BAE40896.1 | |
| AK172230.1 | 2 | 1025 | BAE42896.1 | |
| AK220478.1 | 13 | 2577 | BAD90502.1 | |
| BC005470.1 | 13 | 1631 | AAH05470.1 |
More Information
Set Membership: |
|

Genomic Map