Assay Details
Target Gene Details
Entrez Gene ID: | 236733 |
Gene Name: | ubiquitin specific peptidase 11 |
Gene Aliases: |
6230415D12Rik, mKIAA4085 |
Location: |
Chr.X:20703909-20720539 on Build GRCm38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Usp11 | NM_145628.4 | 21 | 2943 | NP_663603.3 |
| AK031767.1 | 21 | 2922 | BAC27541.1 | |
| AK169116.1 | 13 | 1766 | BAE40896.1 | |
| AK172230.1 | 8 | 1906 | BAE42896.1 | |
| AK220478.1 | 19 | 3458 | BAD90502.1 | |
| BC005470.1 | 19 | 2512 | AAH05470.1 |
More Information
Set Membership: |
|

Genomic Map