Assay Details
Target Gene Details
Entrez Gene ID: | 72691 |
Gene Name: | calcium homeostasis modulator 2 |
Gene Aliases: |
2810048G17Rik, Fam26b |
Location: |
Chr.19:47132232-47138294 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Calhm2 | NM_133746.5 | 1 | 259 | NP_598507.3 |
| AK012923.1 | 1 | 186 | BAC25382.1 | |
| AK040739.1 | 1 | 260 | BAE43310.1 | |
| AK089278.1 | 1 | 260 | BAC40825.1 | |
| AK170068.1 | 1 | 230 | BAE41544.1 | |
| BC019197.1 | 1 | 178 | AAH19197.1 | |
| BC096564.1 | AAH96564.1 |

Genomic Map