Assay Details
Target Gene Details
Entrez Gene ID: | 14704 |
Gene Name: | guanine nucleotide binding protein (G protein), gamma 3 |
Gene Aliases: |
- |
Location: |
Chr.19:8836929-8839246 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gng3 | NM_010316.3 | 3 | 1261 | NP_034446.1 |
| AK032646.1 | 3 | 1209 | BAC27969.1 | |
| BE949484.1 | 1 | 454 |
More Information
Set Membership: |
|

Genomic Map