Assay Details
Target Gene Details
Entrez Gene ID: | 75805 |
Gene Name: | neurolysin (metallopeptidase M3 family) |
Gene Aliases: |
4930472G13Rik, C79345 |
Location: |
Chr.13:104023050-104109660 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nln | NM_029447.2 | 13 | 3439 | NP_083723.1 |
| AK015561.1 | 2 | 1448 | ||
| AK050120.1 | 3 | 1553 | BAC34076.1 | |
| AK079090.1 | 13 | 3440 | BAC37533.1 | |
| AK139150.1 | 1 | 457 | ||
| AK146364.1 | 13 | 3388 | BAE27114.1 | |
| BC016224.1 | 13 | 3412 | AAH16224.1 |
More Information
Set Membership: |
|

Genomic Map