Assay Details
Target Gene Details
Entrez Gene ID: | 22017 |
Gene Name: | thiopurine methyltransferase |
Gene Aliases: |
AW106912 |
Location: |
Chr.13:47021987-47043338 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tpmt | NM_016785.2 | 9 | 1300 | NP_058065.2 |
| AK079074.1 | 2 | 555 |
More Information
Set Membership: |
|

Genomic Map