Assay Details
Target Gene Details
Entrez Gene ID: | 20610 |
Gene Name: | SMT3 suppressor of mif two 3 homolog 3 (yeast) |
Gene Aliases: |
2810014B19Rik, D10Ertd345e, SMT3A, SUMO-3, Smt3h1 |
Location: |
Chr.10:77606097-77618331 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sumo3 | NM_001301671.1 | 4 | 2031 | NP_001288600.1 |
| NM_001301672.1 | 4 | 1863 | NP_001288601.1 | |
| NM_001301673.1 | 4 | 1914 | NP_001288602.1 | |
| NM_019929.4 | 4 | 1905 | NP_064313.1 | |
| NR_125903.1 | 3 | 1634 | ||
| NR_125904.1 | 5 | 2013 | ||
| AK148306.1 | 4 | 1769 | BAE28469.1 |

Genomic Map