Assay Details
Target Gene Details
Entrez Gene ID: | 217666 |
Gene Name: | L-2-hydroxyglutarate dehydrogenase |
Gene Aliases: |
BC016226 |
Location: |
Chr.12:69690436-69724874 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| L2hgdh | NM_145443.2 | 10 | 2861 | NP_663418.1 |
| AK139539.1 | 1 | 1189 | ||
| AK145228.1 | 10 | 2860 | BAE26312.1 | |
| AK168429.1 | 10 | 2809 | BAE40337.1 | |
| BC016226.1 | 10 | 2805 | AAH16226.1 |
More Information
Set Membership: |
|

Genomic Map