Assay Details
Target Gene Details
Entrez Gene ID: | 21665 |
Gene Name: | thymine DNA glycosylase |
Gene Aliases: |
E130317C12Rik, JZA-3, Jza1 |
Location: |
Chr.10:82629247-82650494 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tdg | NM_011561.2 | 10 | 2978 | NP_035691.2 |
| NM_172552.3 | 10 | 3077 | NP_766140.2 | |
| AK053870.1 | 10 | 3005 | BAC35566.1 | |
| AK161792.1 | ||||
| AK164090.1 | BAE37621.1 | |||
| AK169768.1 | BAE41355.1 |
More Information
Set Membership: |
|

Genomic Map