Assay Details
Target Gene Details
Entrez Gene ID: | 624784 |
Gene Name: | thymine DNA glycosylase pseudogene |
Gene Aliases: |
ENSMUSG00000051559 |
Location: |
Chr.10:42052558-42055453 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm9855 | NR_037190.1 | 1 | 58 | |
| AF069519.1 | AAC31900.1 | |||
| AK089915.1 | 1 | 62 | BAC40994.1 |

Genomic Map