Assay Details
Target Gene Details
Entrez Gene ID: | 78802 |
Gene Name: | tetratricopeptide repeat domain 30A1 |
Gene Aliases: |
4930506L13Rik |
Location: |
Chr.2:75979106-75981967 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ttc30a1 | NM_030188.3 | 1 | 126 | NP_084464.3 |
| AK019670.1 | ||||
| BU610816.1 | 1 | 126 |

Genomic Map