Assay Details
Target Gene Details
Entrez Gene ID: | 69942 |
Gene Name: | ring finger protein 113A1 |
Gene Aliases: |
2810428C21Rik, Rnf113a |
Location: |
Chr.X:37191343-37192467 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rnf113a1 | NM_153503.1 | 1 | 1015 | NP_705723.1 |
| AK013178.1 | 1 | 1078 | ||
| AK146283.1 | 1 | 1114 | BAE27041.1 | |
| BC024906.1 | 1 | 1015 | AAH24906.1 |
More Information
Set Membership: |
|

Genomic Map