Assay Details
Target Gene Details
Entrez Gene ID: | 56078 |
Gene Name: | carbonic anhydrase 5b, mitochondrial |
Gene Aliases: |
7330410H16Rik, CAVB, Ca5b, CarVb, D730005F19Rik |
Location: |
Chr.X:163976822-164028010 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Car5b | NM_181315.4 | 8 | 2909 | NP_851832.2 |
| AK041253.1 | 1 | 1551 | ||
| AK163076.1 | 1 | 590 | ||
| CD802657.1 | 1 | 557 |

Genomic Map