Assay Details
Target Gene Details
Entrez Gene ID: | 75705 |
Gene Name: | eukaryotic translation initiation factor 4B |
Gene Aliases: |
2310046H11Rik, AL024095, C85189, Eif4a2 |
Location: |
Chr.15:102073773-102097173 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eif4b | NM_145625.3 | 15 | 2428 | NP_663600.2 |
| AK045250.1 | 15 | 2272 | BAC32280.1 | |
| AK148012.1 | 15 | 2278 | BAE28289.1 | |
| AK161693.1 | 15 | 2261 | BAE36536.1 | |
| AK163266.1 | 15 | 2279 | ||
| AK166363.1 | 15 | 2280 | BAE38733.1 | |
| AK168425.1 | 15 | 2275 | ||
| AK170256.1 | 15 | 2279 | BAE41664.1 | |
| AK170354.1 | 15 | 2261 | BAE41740.1 | |
| AK217495.1 | 1 | 189 | ||
| BC007171.1 | 14 | 2227 | AAH07171.1 |
More Information
Set Membership: |
|

Genomic Map