Assay Details
Target Gene Details
Entrez Gene ID: | 94223 |
Gene Name: | DiGeorge syndrome critical region gene 8 |
Gene Aliases: |
D16H22S1742E, D16H22S788E, D16Wis2, Gy1, N41, Vo59c07, mir-1306 |
Location: |
Chr.16:18253948-18289252 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dgcr8 | NM_033324.2 | 14 | 3366 | NP_201581.2 |
| AB086857.1 | 14 | 3366 | BAD06468.1 | |
| AK086233.1 | 13 | 3558 | ||
| BC062919.1 | 14 | 3386 | AAH62919.1 | |
| BC066118.1 | 14 | 3413 | AAH66118.1 |
Target Gene Details
Entrez Gene ID: | 15547 |
Gene Name: | TRM2 tRNA methyltransferase 2A |
Gene Aliases: |
Htf9c |
Location: |
Chr.16:18248830-18254772 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trmt2a | NM_001080999.2 | NP_001074468.1 | ||
| NM_001081000.2 | NP_001074469.1 | |||
| NM_001195205.1 | NP_001182134.1 | |||
| AK080170.1 | BAC37838.1 |
More Information
Set Membership: |
|

Genomic Map