Assay Details
Target Gene Details
Entrez Gene ID: | 12983 |
Gene Name: | colony stimulating factor 2 receptor, beta, low-affinity (granulocyte-macrophage) |
Gene Aliases: |
AI848964, AIC2B, Bc, CDw131, Csf2rb1, Csfgmrb, Il3r, Il3rb, Il3rb1, Il5rb |
Location: |
Chr.15:78325797-78351001 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Csf2rb | NM_007780.4 | 14 | 4177 | NP_031806.3 |
| AK154286.1 | 14 | 4114 |
More Information
Set Membership: |
|

Genomic Map