Assay Details
Target Gene Details
Entrez Gene ID: | 68338 |
Gene Name: | golgi transport 1 homolog A (S. cerevisiae) |
Gene Aliases: |
0610012C01Rik, Kiss1, metastatin |
Location: |
Chr.1:133309823-133323026 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Golt1a | NM_026680.4 | 5 | 1004 | NP_080956.1 |
| AK141378.1 | 5 | 1005 | BAE24665.1 |
Target Gene Details
Entrez Gene ID: | 280287 |
Gene Name: | KiSS-1 metastasis-suppressor |
Gene Aliases: |
kisspeptin, metastatin |
Location: |
Chr.1:133321887-133329722 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kiss1 |
More Information
Set Membership: |
|

Genomic Map