Assay Details
Target Gene Details
Entrez Gene ID: | 240120 |
Gene Name: | zinc finger protein 119b |
Gene Aliases: |
- |
Location: |
Chr.17:55938373-55956178 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfp119b | NM_146249.4 | 4 | 1754 | NP_666361.1 |
| BC031441.1 | 4 | 1754 | AAH31441.1 | |
| BC137918.1 | ||||
| BC137919.1 |
More Information
Set Membership: |
|

Genomic Map