Assay Details
Target Gene Details
Entrez Gene ID: | 619371 |
Gene Name: | syntaxin-binding protein 3, pseudogene |
Gene Aliases: |
Munc18c(L), Stxbp3-1, Stxbp3b |
Location: |
Chr.19:9557606-9559248 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Stxbp3-ps | NR_073559.1 | 2 | 631 | |
| DQ083989.1 | 2 | 631 | AAY87156.1 |

Genomic Map