Assay Details
Target Gene Details
Entrez Gene ID: | 20437 |
Gene Name: | seven in absentia 1A |
Gene Aliases: |
AA982064, AI853500, Sinh1a |
Location: |
Chr.8:86723920-86746006 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Siah1a | NM_009172.2 | 2 | 1788 | NP_033198.1 |
| AA420282.1 | ||||
| AK019266.1 | 1 | 586 | ||
| BE634662.1 | 1 | 327 | ||
| Z19579.1 | 2 | 1716 | CAA79630.1 |
More Information
Set Membership: |
|

Genomic Map