Assay Details
Target Gene Details
Entrez Gene ID: | 68051 |
Gene Name: | nuclear transport factor 2 |
Gene Aliases: |
2700067I02Rik, AI115459, AI413596, AW546000, Ntf2, Ntf3, PP15 |
Location: |
Chr.8:105860634-105880402 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nutf2 | NM_026532.3 | 5 | 989 | NP_080808.1 |
| AK028410.1 | 5 | 979 | BAC25936.1 | |
| AK188763.1 | 1 | 160 | ||
| BC003955.1 | 5 | 989 | AAH03955.1 | |
| BC086773.1 | 5 | 956 | AAH86773.1 |

Genomic Map