Assay Details
Target Gene Details
Entrez Gene ID: | 94212 |
Gene Name: | phosphoprotein associated with glycosphingolipid microdomains 1 |
Gene Aliases: |
Cbp, F730007C19Rik, Pag |
Location: |
Chr.3:9687479-9833679 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pag1 | NM_001195031.1 | NP_001181960.1 | ||
| NM_053182.5 | NP_444412.2 | |||
| AF250192.1 | AAG44565.1 | |||
| AK040230.1 | BAC30545.1 | |||
| AK076515.1 | ||||
| AK089329.1 | BAC40842.1 | |||
| AK156000.1 | BAE33539.1 | |||
| AK156986.1 | ||||
| AK172106.1 | BAE42829.1 | |||
| BC145328.1 | ||||
| BC145761.1 | ||||
| BC145763.1 |

Genomic Map